missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen Isopropyl Ester, TRC

Product Code. 30402728
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402728 1 g
30408774 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402728 Supplier Toronto Research Chemicals Supplier No. TRCI1400901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64622-17-9
Molecular Formula C16 H24 O2
Molecular Weight (g/mol) 248.36
Synonym Isopropyl 2-(4-Isobutylphenyl)propanoate
IUPAC Name propan-2-yl 2-[4-(2-methylpropyl)phenyl]propanoate
SMILES CC(C)Cc1ccc(cc1)C(C)C(=O)OC(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H24O2/c1-11(2)10-14-6-8-15(9-7-14)13(5)16(17)18-12(3)4/h6-9,11-13H,10H2,1-5H3
Formula Weight 248.1776
Chemical Name or Material Isopropyl 2-(4-Isobutylphenyl)propanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.