missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen Alcohol, TRC

Product Code. 30401502
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401502 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401502 Supplier Toronto Research Chemicals Supplier No. TRCI1400451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2RS)-2-[4-(2-Methylpropyl)-phenyl]propan-1-ol
CAS 36039-36-8
Molecular Formula C13 H20 O
Quantity 1 g
Synonym (2RS)-2-[4-(2-Methylpropyl)-phenyl]propan-1-ol, Ibuprofen Imp. P (EP), Benzeneethanol, β-methyl-4-(2-methylpropyl)-, β-Methyl-4-(2-methylpropyl)benzeneethanol, 2-(4-Isobutylphenyl)-1-propanol, 2-(4-Isobutylphenyl)propanol, 2-(4-Isobutylphenyl)propyl alcohol, 2-[4-(2-Methylpropyl)phenyl]propan-1-ol, Ibuprofen alcohol, (±)-Ibuprofen alcohol
InChI Formula InChI=1S/C13H20O/c1-10(2)8-12-4-6-13(7-5-12)11(3)9-14/h4-7,10-11,14H,8-9H2,1-3H3
SMILES CC(C)Cc1ccc(cc1)C(C)CO
IUPAC Name 2-[4-(2-methylpropyl)phenyl]propan-1-ol
Molecular Weight (g/mol) 192.3
Formula Weight 192.1514
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.