missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen, 99%, Thermo Scientific Chemicals

Product Code. 11964991 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11964991 50 g Glass bottle 50g
15853855 1 g Glass bottle 1g
10768501 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11964991 Supplier Thermo Scientific Acros Supplier No. 258610500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ibuprofen, 2-4-isobutylphenyl propanoic acid, CAS # 15687-27-1, is a laboratory form of the over-the-counter nonsteroidal anti-inflammatory (NSAID) drug.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Ibuprofen (2-4-isobutylphenyl propanoic acid) is a common NSAID that reduces pyrogen release and inflammation
  • Ibuprofen is a racemic mixture that consists of an R-enantiomer and S-enantiomer; the S-enantiomer is proposed to be the stronger acting agent
  • It is a propionic acid derivative, a short-chain saturated fatty acid bound to ethane
  • Ibuprofen inhibits cyclooxygenases and reduces prostaglandin formation, inhibiting fever and pain

Applications

  • Ibuprofen reduces both local and systemic inflammation; suitable for use in vitro or in vivo to study specific or broad mechanisms
  • Intended for sensitive biochemical applications such as HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15687-27-1
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.29
MDL Number MFCD00010393
InChI Key HEFNNWSXXWATRW-UHFFFAOYNA-N
PubChem CID 3672
ChEBI CHEBI:5855
IUPAC Name 2-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)CC1=CC=C(C=C1)C(C)C(O)=O

Specifications

Melting Point 75.0°C to 78.0°C
Color White
Boiling Point 157.0°C (4.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2CHCH2C6H4CH(CH3)COOH
Quantity 50 g
Merck Index 15,4919
Synonym ibuprofen, 2-4-isobutylphenyl propanoic acid, motrin, brufen, nurofen, advil, dolgit, liptan, nuprin, anflagen
Solubility Information Solubility in water: insoluble. Other solubilities: readily soluble in most organic solvents
Formula Weight 206.27
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Ibuprofen
Show More Show Less
compliance-icons
Product Identifier
  • Ibuprofen
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.