missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen 2-Monoglyceride, TRC

Product Code. 30397947
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397947 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397947 Supplier Toronto Research Chemicals Supplier No. TRCI400140250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Dihydroxyprop-2-yl 2-(4-Isobutylphenyl)propanoate
CAS 124635-85-4
Molecular Formula C16 H24 O4
Quantity 250 mg
Synonym 1,3-Dihydroxyprop-2-yl 2-(4-Isobutylphenyl)propanoate
InChI Formula InChI=1S/C16H24O4/c1-11(2)8-13-4-6-14(7-5-13)12(3)16(19)20-15(9-17)10-18/h4-7,11-12,15,17-18H,8-10H2,1-3H3
SMILES CC(C)Cc1ccc(cc1)C(C)C(=O)OC(CO)CO
IUPAC Name 1,3-dihydroxypropan-2-yl 2-[4-(2-methylpropyl)phenyl]propanoate
Molecular Weight (g/mol) 280.359
Formula Weight 280.167
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.