missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibufenac-13C6, TRC
Specifications
Specifications
| Chemical Name or Material | Ibufenac-13C6 |
| Molecular Formula | 13C6 C6 H16 O2 |
| Quantity | 10 mg |
| Synonym | 4-(2-Methylpropyl)benzeneacetic Acid-13C6, 4-(2-Methylpropyl)phenylacetic Acid-13C6, 4-Isobutoxyphenylacetic Acid-13C6, 4-Isobutylphenylacetic Acid-13C6, Dytransin-13C6, Ibunac-13C6, Medirex-13C6, NSC 99976-13C6 |
| InChI Formula | InChI=1S/C12H16O2/c1-9(2)7-10-3-5-11(6-4-10)8-12(13)14/h3-6,9H,7-8H2,1-2H3,(H,13,14)/i3+1,4+1,5+1,6+1,10+1,11+1 |
| SMILES | CC(C)C[13c]1[13cH][13cH][13c](CC(=O)O)[13cH][13cH]1 |
| IUPAC Name | 2-[4-(2-methylpropyl)(1,2,3,4,5,6-13C6)cyclohexa-1,3,5-trien-1-yl]acetic acid |
| Molecular Weight (g/mol) | 198.21 |
| Formula Weight | 198.135 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?