missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Format | Powder |
| For Use With (Equipment) | Expedite Synthesizers |
| Product Type | iBu-rG CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | Guanosine (G) |
| Shipping Condition | Room Temperature |
| Purity | 99% |
| Quantity | 1 g |
| Reagent Type | Phosphoramidite |
| Grade | TheraPure™ |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?