missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IBTK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561733
This item is not returnable.
View return policy
Beskrivning
Immunogen sequence: ILFVTQDGEG FRGRWFEEKR KSSEKKEILS NLHNSSSDVS YVSDINSVYE RIRLEKLTFA HRAVSVSTDP SGCNFAILQS DPKTSLYEIP AVSSSSFFEE FGKLLREADE MDSIHDVTFQ VGN Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
The protein encoded by this gene binds to Bruton's tyrosine kinase (BTK) and downregulates BTK's kinase activity. In addition, the encoded protein disrupts BTK-mediated calcium mobilization and negatively regulates the activation of nuclear factor-kappa-B-driven transcription.
Specifikationer
| IBTK | |
| Polyclonal | |
| Unconjugated | |
| IBTK | |
| Bruton agammaglobulinemia tyrosine kinase inhibitor; BTBD26; BTK-binding protein; BTKI; IBTK; Inhibitor of Bruton tyrosine kinase; inhibitor of Bruton's tyrosine kinase-alpha; inhibitor of Bruton's tyrosine kinase-beta; inhibitor of Bruton's tyrosine kinase-gamma; KIAA1417; RP1-93K22.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25998 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P2D0 | |
| IBTK | |
| Recombinant protein corresponding to Human IBTK. Recombinant protein control fragment (Product #RP-103025). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering