missing translation for 'onlineSavingsMsg'
Learn More

IBTK Antibody, DyLight 650, Novus Biologicals™

Product Code. 18425873 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425873 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425873 Supplier Novus Biologicals Supplier No. NBP150033C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IBTK Polyclonal antibody specifically detects IBTK in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IBTK
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Bruton agammaglobulinemia tyrosine kinase inhibitor, BTK-binding protein, BTKI, DKFZP564B116, IBtk, inhibitor of Bruton agammaglobulinemia tyrosine kinase, inhibitor of Bruton tyrosine kinase, inhibitor of Bruton's tyrosine kinase, inhibitor of Bruton's tyrosine kinase-alpha, inhibitor of Bruton's tyrosine kinase-beta, inhibitor of Bruton's tyrosine kinase-gamma, KIAA1417, MGC142256, MGC142258, RP1-93K22.1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1050 and 1100 of human Inhibitor of Bruton Agammaglobulinemia Tyrosine Kinase, using the numbering given in entry NP_056340.2 (GeneID 25998).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25998
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.