missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IBSP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541327
This item is not returnable.
View return policy
Description
Peptide sequence: SATTLGYGED ATPGTGYTGL AAIQLPKKAG DITNKATKEK ESDEEEEEEE Sequence homology: Dog: 79%; Human: 100%.
The protein encoded by this gene is a major structural protein of the bone matrix. It constitutes approximately 12% of the noncollagenous proteins in human bone and is synthesized by skeletal-associated cell types, including hypertrophic chondrocytes, osteoblasts, osteocytes, and osteoclasts. The only extraskeletal site of its synthesis is the trophoblast. This protein binds to calcium and hydroxyapatite via its acidic amino acid clusters, and mediates cell attachment through an RGD sequence that recognizes the vitronectin receptor.
Specifications
| IBSP | |
| Polyclonal | |
| Unconjugated | |
| Ibsp | |
| BNSP; Bone sialoprotein 2; bone sialoprotein II; Bsp; BSP II; Bsp2; BSPII; BSP-II; Cell-binding sialoprotein; IBSP; integrin binding sialoprotein; integrin-binding sialoprotein; SP-II | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3381 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P21815 | |
| Ibsp | |
| Synthetic peptide directed towards the N-terminal of human IBSP (aa 108-157). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction