Learn More
DiscoverX Ibrutinib (PCI-32765)
Product Code. 17362559
Encompass
Click to view available options
:
5 mg
Product Code. 17362559
Brand: DiscoverX 921243
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ibrutinib (PCI-32765) is an inhibitor of BTK, YES, and other SRC tyrosine kinases. Provided as 5mg of dry powder.
Ibrutinib (PCI-32765, Imbruvica) is an FDA approved drug that targets B-cell malignancies and is approved for treatment of mantle cell lymphoma, chronic lymphocytic leukemia, and Waldenstrom's macroglobulinemia. Ibrutinib is an orally-bioavailable compound that is a potent and selective inhibitor of Bruton's tyrosine kinase (BTK). Also inhibits YES and other SRC family tyrosine kinases. Provided as 5mg of dry powder.
Specifications
Quantity 5 mg