missing translation for 'onlineSavingsMsg'
Learn More

Ibrutinib, MedChemExpress

Product Code. p-200096745
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16474338

Brand: Medchem Express HY10997

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ibrutinib (PCI-32765) is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.

Ibrutinib (PCI-32765) is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.

Specifications

Chemical Name or Material Ibrutinib
CAS 936563-96-1
Color White
Purity Grade Notes Research
Molecular Formula C25H24N6O2
Quantity 10 mg
Synonym PCI-32765
Solubility Information DMSO : 100 mg/mL (227.01 mM; Need ultrasonic)
SMILES C=CC(N1C[C@H](N2N=C(C3=CC=C(OC4=CC=CC=C4)C=C3)C5=C(N)N=CN=C52)CCC1)=O
Molecular Weight (g/mol) 440.5
Formula Weight 440.5
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.