missing translation for 'onlineSavingsMsg'
Learn More

Ibrutinib-biotin, MedChemExpress

Product Code. 16449526
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449526 50 mg 50mg
16445697 5 mg 5mg
16422958 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16449526 Supplier Medchem Express Supplier No. HY100342
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ibrutinib-biotin is a probe that consists of Ibrutinib linked to biotin via a long chain linker, extracted from patent WO2014059368A1 Compound 1-5, has an IC50 of 0.755-1.02 nM for BTK.

Ibrutinib-biotin is a probe that consists of Ibrutinib linked to biotin via a long chain linker, extracted from patent WO2014059368A1 Compound 1-5, has an IC50 of 0.755-1.02 nM for BTK.

Specifications

Chemical Name or Material Ibrutinib-biotin
CAS 1599432-18-4
Color Colorless
Purity Grade Notes Research
Molecular Formula C56H80N12O9S
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (45.56 mM; Need ultrasonic)
SMILES O=C(CCCC(NCCCOCCOCCOCCCNC(CCCC[C@@H]1SC[C@H](N2)[C@@H]1NC2=O)=O)=O)NCCCCN(C)C/C=C/C(N3CCC[C@@H](N4N=C(C5=CC=C(OC6=CC=CC=C6)C=C5)C7=C(N)N=CN=C74)C3)=O
Molecular Weight (g/mol) 1097.37
Formula Weight 1097.37
Percent Purity 98.04%
Grade Research
Physical Form Oil
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.