missing translation for 'onlineSavingsMsg'
Learn More

Ibotenic acid, MedChemExpress

Product Code. 16479278
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479278 10 mg 10mg
16474937 50 mg 50mg
16471708 5 mg 5mg
16433956 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16479278 Supplier Medchem Express Supplier No. HYN2311
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ibotenic acid has agonist activity at both the N-methyl-D-aspartate (NMDA) and trans-ACPD or metabolotropic quisqualate (Qm) receptor sites.

Ibotenic acid has agonist activity at both the N-methyl-D-aspartate (NMDA) and trans-ACPD or metabolotropic quisqualate (Qm) receptor sites.

Specifications

Chemical Name or Material Ibotenic acid
CAS 2552-55-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C5H6N2O4
Quantity 10 mg
Synonym (RS)-Ibotenic acid DL-Ibotenic acid
Solubility Information H2O : 5 mg/mL (31.62 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (31.62 mM; Need ultrasonic)
SMILES O=C(O)C(N)C(ON1)=CC1=O
Molecular Weight (g/mol) 158.11
Formula Weight 158.11
Percent Purity 98.27%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.