missing translation for 'onlineSavingsMsg'
Learn More

IBMX, MedChemExpress

Product Code. 16431327
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16431327 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16431327 Supplier Medchem Express Supplier No. HY12318
 Restricted Item
Estimated Shipment: 08-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress IBMX is a broad-spectrum phosphodiesterase (PDE) inhibitor, with IC50s of 6.5, 26.3 and 31.7 μM for PDE3, PDE4 and PDE5, respectively.

IBMX is a broad-spectrum phosphodiesterase (PDE) inhibitor, with IC50s of 6.5, 26.3 and 31.7 μM for PDE3, PDE4 and PDE5, respectively.

Specifications

Chemical Name or Material IBMX
Physical Form Solid
CAS 28822-58-4
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C10H14N4O2
For Use With (Application) COVID-19-immunoregulation
Synonym 3-Isobutyl-1-methylxanthine Isobutylmethylxanthine
Solubility Information DMSO : 50 mg/mL (224.98 mM; Need ultrasonic) ∣Ethanol : ≥ 7.14 mg/mL (32.13 mM)
SMILES O=C(N1C)N(CC(C)C)C2=C(N=CN2)C1=O
Molecular Weight (g/mol) 222.24
Formula Weight 222.24
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.