missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBlot™ 3 Transfer Stacks, mini, low-fluorescence PVDF

Product Code. 30484883
Change view
Click to view available options
Quantity:
10 Stacks
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30484883 10 Stacks Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30484883 Supplier Invitrogen™ Supplier No. IB34004
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Invitrogen Low Fluorescence PVDF Membranes exhibit exceptionally low autofluorescence across all commonly used fluorescent imaging channels. Low fluorescent background enhances signal-to-noise ratios, resulting in more sensitive, higher quality and more reliable western blot data.

Invitrogen iBlot 3 Transfer Stacks are used with the iBlot 3 Western Blot Transfer Device to transfer proteins from an SDS-PAGE gel to a membrane when performing western blotting experiments. These pre-assembled, single-use consumables utilize copper-fiber electrode sheets and optimized cathode and anode solid buffer gels to eliminate several sources of variability—helping ensure consistent and repeatable results compared with classical methods.

The Low Fluorescence PVDF membranes exhibit exceptionally low autofluorescence across all commonly used fluorescent imaging channels. Low fluorescent background enhances signal-to-noise ratios, resulting in more sensitive, higher quality, and more reliable western blot data.

iBlot 3 Transfer Stacks allow you to:

  • Boost signal-to-noise-ratio—low autofluorescence enhances signal clarity
  • Expand multiplexing capability—low fluorescent background across channels broadens fluorophore options allowing users to detect more targets or minimize spectral cross-talk
  • Minimize variability—single-use copper electrodes prevent salt build-up from impacting results over time
  • Expedite setup—pre-assembled transfer stacks contain solid gel ion matrix, which replaces traditional transfer buffer
  • Simplify clean up—transfer stack is self-contained within a disposable tray, simplifying cleanup to one step (wiping the blotting surface of the transfer device)

Compatibility

Although optimized for fluorescent detection, Low Fluorescence PVDF membranes perform similarly to standard PVDF when used in chemiluminescent detection.

All iBlot 3 transfer stacks are compatible with common commercially available protein gels, including Bis-Tris, Tris-Glycine, Tris-Acetate, and Tricine chemistries. They are intended for use on the iBlot 3 Transfer Device only.

Use

Mini-size transfer stacks (8 x 8 cm) can be used with one mini-size gel. To enable optimal performance in fluorescent applications, Low Fluorescence PVDF membranes are provided dry, in a separate pouch within the box. Activate the membrane before use by soaking in 100% methanol or ethanol, followed by a quick rinse with distilled water. Once activated, simply assemble the transfer stack, choose a transfer method, and press Go.

The iBlot 3 Western Blot Transfer System

The iBlot 3 Western Blot Transfer System, which includes iBlot 3 Transfer Stacks and the iBlot 3 Western Blot Transfer Device, utilizes dry blotting technology to produce higher quality and more reliable western blot data while reducing protein transfer to 3–8 minutes.

The iBlot 3 system features two independently controlled transfer stations, allowing the device to be shared with coworkers and simultaneously running up to four mini gels or two midi gels with two different programs. In addition, the robust design and adjustable cooling function reduces heat buildup, facilitating better run-to-run consistency.

How dry blotting technology works

Buffer ion reservoirs are incorporated into the gel matrix of transfer stacks instead of buffer tanks or soaked filter papers. The high density of ions in the gel matrix enables rapid protein transfer. During blotting, the copper anode does not generate oxygen gas as a result of water electrolysis, reducing blot distortion. (Conventional protein transfer techniques, including wet, semi-wet, and semi-dry, use inert electrodes that generate oxygen.)

With the iBlot dry blotting system, transfer time is reduced by the shortened distance between electrodes, high field strength, and high current. Trapped air bubbles, often created during the manual preparation of the blotting sandwich layers, are easy to avoid due to our exceptional de-bubbling design that promotes even and complete transfer. With the iBlot 3 system, there is no need to prepare buffers or clean up after blotting. The total preparation and run time is normally less than ten minutes per blot.

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 8 cm
Width (Metric) 8 cm
For Use With (Application) Western Blotting
For Use With (Equipment) iBlot 3
Quantity 10 Stacks
Format Transfer Stack
Material PVDF
Pore Size 0.3 μm
Content And Storage 10 transfer stacks, 10 Low Fluorescence PVDF Membranes, 10 Absorbent Pads, 10 Filter Papers; Store at room temperature
Dimensions (LxW) 8 x 8 cm (Mini)
Product Line iBlot
Sufficient For 10 Blots
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.