Learn More
Nitrotryptophan Monoclonal Antibody (4F8), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA545821
Description
A 1:1,000 dilution of MA5-45821 was sufficient for detection of 6-Nitrotryptophan in 1 μg of 6-Nitrotyrptophan conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody.∣Specific for 6-Nitrotyrptophan modified proteins. Doss not cross-react with 7-Ketocholoesterol modified proteins.
6-nitrotryptophan (6-NO2-Trp) is shown to be one of the most stable and major ONOO- modified amino acids, and growing evidences suggest that nitrotryptophan may play a significant role in the cellular regulation under nitrosative stress. Reactive nitrogen species (RNS) are formed in a variety of diseases such as atherosclerosis, inflammation, cancer and neurodegenerative disiases. Peroxinitrite (ONOO-) is known as a potent biological oxidant, which is formed via very fast reaction between nitric oxide (NO) and superoxide (O2-). Peroxinitrite can induce nitration of aromatic amino acids such as Tyr, Trp, Phe and His, which results in modulation of protein functions.
Specifications
| Nitrotryptophan | |
| Monoclonal | |
| 1 mg/mL | |
| 2.15mM MES, 47.35mM phosphate with 1.033mM EDTA and no preservative; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Liquid | |
| Chemical |
| ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
| 4F8 | |
| APC | |
| 6-Nitrotryptophan; 6-NO2-Trp; Nitrotryptophan; NW | |
| Synthetic 6-Nitrotryptophan modified Keyhole Limpet Hemocyanin (KLH). | |
| 100 μg | |
| Primary | |
| 4° C | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.