missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBind™ Flex Western Starter Kit, fluorescent detection

Product Code. 17089843
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17089843

Brand: Invitrogen™ SLF2002S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The iBind Flex Western Starter Kit for fluorescent detection includes an iBind Flex Western Device , iBind Flex Cards, and an iBind Flex Fluorescent Detection (FD) Solution Kit.

The iBind Flex Western Device is a small benchtop device that automates western blot processing (immunodetection). It applies mechanical pressure to an iBind Flex Card to generate sequential lateral flow (SLF) of immunodetection solutions for the blocking, antibody binding, and washing steps. The glass fiber matrix of the iBind Flex Card allows for homogenous and consistent flow of the solutions over the membrane, increasing the antigen-antibody interaction.

The iBind Flex Western Device allows all solutions to be prepared and loaded in the device at the start of the procedure; all subsequent steps proceed automatically through SLF action—no electricity or batteries required. Blots are ready for the final detection steps in approximately 2.5 hours.

The iBind Flex Western Device includes three user-interchangeable well inserts to process up to one midi blot, two mini blots, or six vertically-cut strip blots when used with iBind Flex Cards, and the iBind Flex Solution Kit or iBind Flex Fluorescent Detection (FD) Solution Kit to process one mini-blot.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 iBind Flex Western Device, 10 iBind Flex Cards (store at room temperature), 1 iBind Flex Fluorescent Detection (FD) Solution Kit (store at 2 to 8°C)
Capacity 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Detection Method Fluorescence
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line iBind
Running Time ∼2.5 hr.
Throughput 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Type Starter Kit
Quantity 1 kit
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.