missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBind Flex PLUS Solution Kit

Product Code. 31921187
Change view
Click to view available options
Quantity:
1 Kit
4 Kits
Unit Size:
4 sets
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31921187 1 Kit Each
31935003 4 Kits 4 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31921187 Supplier Invitrogen™ Supplier No. SLF2030
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The iBind Flex PLUS Solution Kit increases signal sensitivity, requires less antibody per run, and simplifies western blotting immunoprocessing when used with the iBind Flex Western System. The iBind Flex PLUS Solution Kit comes with two ready to use solutions, iBind Flex PLUS Solution and iBind Flex PLUS Antibody Diluent.

  • The iBind Flex PLUS Solution comes ready to use with no preparation required. It is used to prewet the iBind Flex card prior to the run and acts as the block and wash buffer for the membrane processing, helping wash away unbound primary and secondary antibodies.
  • The iBind Flex PLUS Antibody Diluent is used to dilute the primary and secondary antibodies. In most experiments, the primary antibody can be diluted to the same recommended concentration as used for traditional blotting, however only 2 mL (mini blot), 4 mL (midi blot) or 0.7 mL (strips) of prepared primary antibody is required with iBind Flex, saving up to 80% of primary antibody consumption. The iBind Flex PLUS Antibody Diluent is specially formulated with bulking agents and stabilizing molecules to improve antibody binding with the target antigen to increase signal intensity.

During a run on the iBind Flex, each solution is released from the iBind Flex device wells through sequential lateral flow (SLF) and are wicked over the iBind Flex card towards the stack region, incubating the membrane with each reagent. The glass fiber matrix of the iBind Flex card allows for homogenous and consistent flow of the solutions to the membrane, increasing the antigen-antibody interaction.

The iBind Flex PLUS Solution Kit, together with the iBind Flex Western Device (Cat. No. SLF2000) and iBind Flex Cards (Cat. No. SLF2010), bring automation and simplicity to western blotting with efficient and hands-free immunoprocessing in as little as 3 hours. Compared to manual methods, the iBind Flex western system requires less primary antibody, enables reproducible results, and is compatible with downstream detection protocols.

Each iBind Flex PLUS Solution Kit contains sufficient reagents for 10 immunoprocessing reactions with flexibility of each reaction supporting 1 midi blot, 2 mini blots, or 6 vertically cut strips.

For iBind Devices, use the iBind PLUS Solution Kit. For fluorescent applications, use the iBind Fluorescent Detection (FD) Solution Kit (Cat. No. SLF1019) or iBind Flex Fluorescent Detection (FD) Solution Kit (Cat. No. SLF2019).

TRUSTED_SUSTAINABILITY

Specifications

Product Type Flex Solution Kit
Content And Storage iBind Flex PLUS Solution, iBind Flex PLUS Antibody Diluent
Store at 2°C to 8°C
For Use With (Equipment) iBind Flex Western Device
No. of Reactions 10 Midi Blots or 20 Mini Blots
Detection Method Colorimetric, Chromogenic, Chemiluminescence
Gel Compatibility Mini Precast Protein Gels, Midi Precast Protein Gels
Membrane Compatibility Nitrocellulose, PVDF
Product Line iBind
Quantity 1 Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.