Learn More
Invitrogen™ iBind™ Flex Midi Insert Replacement
Description
The iBind™ Flex Midi Insert Replacement is an accessory for the iBind™ Flex Western Device (Cat. No. SLF2000) to process one midi blot.
iBind Flex Western System
The iBind™ Flex Western System offers automated western blot processing and can accommodate various blot formats including midi, mini, and vertically cut strips. Based on Sequential Lateral Flow (SLF) technology, all blocking, antibody incubation and washing steps during immunodetection are hands-free. Compared to manual methods, the iBind Flex Western System requires less primary antibody, enables reproducible results and is compatible with downstream detection protocols.
Specifications
Specifications
| Product Type | Replacement Well Inserts |
| Description | Midi well insert with single well on row 1 and 3 |
| Kit Contents | Single well on row 1 and 3 |
| For Use With (Application) | Western Blotting |
| For Use With (Equipment) | iBind |
| Product Line | iBind |
| Type | Replacement Insert |
| Quantity | 1 insert |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.