missing translation for 'onlineSavingsMsg'
Learn More

Iberdomide, MedChemExpress

Product Code. 16453328
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453328

Brand: Medchem Express HY101291

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Iberdomide (CC-220) is an orally active and potent cereblon (CRBN) E3 ligase modulator (CELMoD) with an IC50 of ∼150 nM for cereblon-binding affinity. Iberdomide, a derivative of Thalidomide (HY-14658), has antitumor and immunostimulatory activities.

Iberdomide (CC-220) is an orally active and potent cereblon (CRBN) E3 ligase modulator (CELMoD) with an IC50 of ∼150 nM for cereblon-binding affinity. Iberdomide, a derivative of Thalidomide (HY-14658), has antitumor and immunostimulatory activities.

Specifications

Chemical Name or Material Iberdomide
CAS 1323403-33-3
Color White
Purity Grade Notes Research
Molecular Formula C25H27N3O5
Quantity 10 mg
Synonym CC-220
Solubility Information DMSO : 125 mg/mL (278.09 mM; Need ultrasonic)
SMILES O=C([C@@H](N(CC1=C2C=CC=C1OCC3=CC=C(CN4CCOCC4)C=C3)C2=O)CC5)NC5=O
Molecular Weight (g/mol) 449.5
Formula Weight 449.5
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.