missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibandronate Sodium Monohydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Ibandronate Sodium Monohydrate |
| CAS | 138926-19-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C9H22NO7P2.Na.H2O |
| Quantity | 1 g |
| Synonym | Phosphonic acid, P,P'-[1-hydroxy-3-(methylpentylamino)propylidene]bis-, sodium salt, hydrate (1:1:1) (ACI), Phosphonic acid, [1-hydroxy-3-(methylpentylamino)propylidene]bis-, monosodium salt, monohydrate (9CI), BM 21.0955Na.H2O, Ibandronate monosodium monohydrate, Ibandronate sodium monohydrate, Monosodium ibandronate monohydrate, Sodium ibandronate monohydrate, P,P'-[1-Hydroxy-3-(methylpentylamino)propylidene]bisphosphonic acid sodium salt hydrate (1:1:1), [1-Hydroxy-3-(methylpentylamino)propylidene]bisphosphonic acid monosodium salt monohydrate |
| InChI Formula | InChI=1S/C9H23NO7P2.Na.H2O/c1-3-4-5-7-10(2)8-6-9(11,18(12,13)14)19(15,16)17;;/h11H,3-8H2,1-2H3,(H2,12,13,14)(H2,15,16,17);;1H2/q;+1;/p-1 |
| SMILES | O.[Na+].CCCCCN(C)CCC(O)(P(=O)(O)O)P(=O)(O)[O-] |
| IUPAC Name | sodium;hydroxy-[1-hydroxy-3-[methyl(pentyl)amino]-1-phosphonopropyl]phosphinate;hydrate |
| Molecular Weight (g/mol) | 359.23 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?