missing translation for 'onlineSavingsMsg'
Learn More

Ibandronate Sodium Monohydrate, TRC

Product Code. 30399163
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399163 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399163 Supplier Toronto Research Chemicals Supplier No. TRCI12000150MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ibandronate Sodium Monohydrate
CAS 138926-19-9
Purity Grade Notes HPLC
Molecular Formula C9H22NO7P2.Na.H2O
Quantity 50 mg
Synonym Phosphonic acid, P,P'-[1-hydroxy-3-(methylpentylamino)propylidene]bis-, sodium salt, hydrate (1:1:1) (ACI), Phosphonic acid, [1-hydroxy-3-(methylpentylamino)propylidene]bis-, monosodium salt, monohydrate (9CI), BM 21.0955Na.H2O, Ibandronate monosodium monohydrate, Ibandronate sodium monohydrate, Monosodium ibandronate monohydrate, Sodium ibandronate monohydrate, P,P'-[1-Hydroxy-3-(methylpentylamino)propylidene]bisphosphonic acid sodium salt hydrate (1:1:1), [1-Hydroxy-3-(methylpentylamino)propylidene]bisphosphonic acid monosodium salt monohydrate
InChI Formula InChI=1S/C9H23NO7P2.Na.H2O/c1-3-4-5-7-10(2)8-6-9(11,18(12,13)14)19(15,16)17;;/h11H,3-8H2,1-2H3,(H2,12,13,14)(H2,15,16,17);;1H2/q;+1;/p-1
SMILES O.[Na+].CCCCCN(C)CCC(O)(P(=O)(O)O)P(=O)(O)[O-]
IUPAC Name sodium;hydroxy-[1-hydroxy-3-[methyl(pentyl)amino]-1-phosphonopropyl]phosphinate;hydrate
Molecular Weight (g/mol) 359.23
Formula Weight 359.0875
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.