missing translation for 'onlineSavingsMsg'
Learn More

Ibandronate Sodium Monohydrate, MedChemExpress

Product Code. 16478346
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16478346

Brand: Medchem Express HYB0515

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ibandronate Sodium Monohydrate is a highly potent nitrogen-containing bisphosphonate used for the treatment of osteoporosis.

Ibandronate Sodium Monohydrate is a highly potent nitrogen-containing bisphosphonate used for the treatment of osteoporosis.

Specifications

Chemical Name or Material Ibandronate Sodium Monohydrate
Physical Form Powder
CAS 138926-19-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H24NNaO8P2
For Use With (Application) Cancer-programmed cell death
Synonym BM-210955 RPR-102289A
Solubility Information H2O : 25 mg/mL (69.59 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES OC(P(O)(O)=O)(P(O)(O[Na])=O)CCN(C)CCCCC.O
Molecular Weight (g/mol) 359.23
Formula Weight 359.23
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.