missing translation for 'onlineSavingsMsg'
Learn More

IBA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16807603 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16807603 20 μL 20µL
16897593 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16807603 Supplier Proteintech Supplier No. 109041AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AIF1 is induced by cytokines and interferon. It is thought to be involved in the negative regulation of growth of vascular smooth muscle cells, which contributes to the anti-inflammatory response to vessel wall trauma.

Specifications

Antigen IBA1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene AIF1
Gene Accession No. O70200, P55008
Gene Alias AIF 1, AIF1, G1, IBA1, IRT 1, Protein G1
Gene Symbols AIF1
Host Species Rabbit
Immunogen IBA1 Fusion Protein Ag1363
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11629, 199
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.