missing translation for 'onlineSavingsMsg'
Learn More

IAXO-102, MedChemExpress

Product Code. p-200098795
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16455906

Brand: Medchem Express HY125171

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IAXO-102 is a TLR4 antagonist which negatively regulates TLR4 signalling. IAXO-102 inhibits MAPK and p65 NF-κB phosphorylation and expression of TLR4 dependent proinflammatory protein. IAXO-102 also prevents experimental abdominal aortic aneurysm development.

IAXO-102 is a TLR4 antagonist which negatively regulates TLR4 signalling. IAXO-102 inhibits MAPK and p65 NF-κB phosphorylation and expression of TLR4 dependent proinflammatory protein. IAXO-102 also prevents experimental abdominal aortic aneurysm development.

Specifications

Chemical Name or Material IAXO-102
Physical Form Solid
CAS 1115270-63-7
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C35H71NO5
For Use With (Application) COVID-19-immunoregulation
Solubility Information Ethanol : 50 mg/mL (85.33 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (8.53 mM; Need ultrasonic)
SMILES CCCCCCCCCCCCCCO[C@@H]1[C@H]([C@@H]([C@@H](CN)O[C@@H]1OC)O)OCCCCCCCCCCCCCC
Molecular Weight (g/mol) 585.94
Formula Weight 585.94
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.