missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iASPP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48732
This item is not returnable.
View return policy
Description
iASPP Polyclonal antibody specifically detects iASPP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Specifications
| iASPP | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockdown Validated | |
| IASPPinhibitor of apoptosis stimulating protein of p53, Inhibitor of ASPP protein, NFkB-interacting protein 1, NKIP1, PPP1R13BL, PPP1R13B-like protein, Protein iASPP, protein phosphatase 1, regulatory (inhibitor) subunit 13 like, protein phosphatase 1, regulatory subunit 13 like, RAINFkB interacting protein 1, relA-associated inhibitor | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DSEAFQSARDFLDMNFQSLAMKHMDLKQMELDTAAAKVDELTKQLESLWSDSPAPPGPQAGPPSRPPRYSSSSIPEPFGSRGSPRKAATDGADTPFGRSESAPTLHPYSP | |
| 0.1 mL | |
| Apoptosis, Cancer, Caspases | |
| 10848 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction