missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IARS2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA555117

Product Code. 15753041

  • £487.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: VASQHNLPMD CLVDEDGVFT DVAGPELQNK AVLEEGTDVV IKMLQTAKNL LKEEKLVHSY PYDWRTKKPV VIRASKQWFI NITDIKTA Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.

The Trip230 gene encodes a protein of 1,978 amino acids with an estimated molecular mass of 230 kDa. The gene is located on chromosome 14q31, which is in a region associated with genetic alterations and thyroid hormone response abnormalities. The Trip230 protein interacts with Rb and thyroid hormone receptor (TR) using two distinct domains. Complex formation with Rb is in a hormone independent manner, but when Trip230 forms a complex with TR, it is in a thyroid hormone-dependent manner. After T3 treatment, Trip230 is phosphorylated and transported to the nucleus, suggesting that phosphorylation may be important for its nuclear translocation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IARS2
Polyclonal
Unconjugated
IARS2
2010002H18Rik; C79125; CAGSSS; EC 6.1.1.5; Iars2; IleRS; isoleucine tRNA ligase 2, mitochondrial; isoleucine--tRNA ligase, mitochondrial; isoleucine-tRNA synthetase 2, mitochondrial; Isoleucyl-tRNA synthetase; isoleucyl-tRNA synthetase 2, mitochondrial; isoleucyl-tRNA synthetase, mitochondrial; mitochondrial isoleucine tRNA synthetase; RGD1311857
Rabbit
Antigen affinity chromatography
RUO
55699
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NSE4
IARS2
Recombinant protein corresponding to Human IARS2. Recombinant protein control fragment (Product #RP-94007).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.