missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IARS Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26942-1-AP-150UL
This item is not returnable.
View return policy
Description
IARS is a member of aminoacyl-tRNA synthetases which catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAS, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Isoleucine-tRNA synthetase belongs to the class-I aminoacyl-tRNA synthetase family and has been identified as a target of autoantibodies in the autoimmune disease polymyositis/dermatomyositis. Two alternatively spliced variants have been isolated that represent alternate 5′ UTRs.Specifications
| IARS | |
| Polyclonal | |
| Unconjugated | |
| IARS | |
| IARS, IARS1, IleRS, ILRS, IRS, Isoleucine tRNA ligase, isoleucyl tRNA synthetase, PRO0785 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3376 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P41252 | |
| IARS | |
| IARS Fusion Protein Ag25686 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction