missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IAH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17808-1-AP-150UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
| IAH1 | |
| Polyclonal | |
| Unconjugated | |
| IAH1 | |
| IAH1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 285148, 298917, 67732 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q2TAA2, Q711G3, Q9DB29 | |
| Iah1 | |
| IAH1 Fusion Protein Ag12089 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction