missing translation for 'onlineSavingsMsg'
Learn More

IACS-010759 hydrochloride, MedChemExpress

Product Code. 16413126
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16413126 25 mg 25mg
16495396 100 mg 100mg
16474548 10 mg 10mg
16447267 5 mg 5mg
16400857 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16413126 Supplier Medchem Express Supplier No. HY112037A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IACS-010759 hydrochlorideis an orally active, potent mitochondrial complex I of oxidative phosphorylation (OXPHOS) inhibitor. IACS-010759 hydrochlorideinhibits proliferation and induces apoptosis in models of brain cancer and acute myeloid leukemia (AML) reliant on OXPHOS. IACS-010759 hydrochloride has the potential for relapsed/refractory AML and solid tumors research.

IACS-010759 hydrochlorideis an orally active, potent mitochondrial complex I of oxidative phosphorylation (OXPHOS) inhibitor. IACS-010759 hydrochlorideinhibits proliferation and induces apoptosis in models of brain cancer and acute myeloid leukemia (AML) reliant on OXPHOS. IACS-010759 hydrochloride has the potential for relapsed/refractory AML and solid tumors research.

Specifications

Chemical Name or Material IACS-010759 hydrochloride
CAS 1807523-99-4
Color White
Purity Grade Notes Research
Molecular Formula C25H26ClF3N6O4S
Quantity 25 mg
Synonym IACS-10759 hydrochloride
Solubility Information DMSO : 60 mg/mL (100.16 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES FC(F)(F)OC1=CC=C(C2=NOC(C3=NN(CC4=CC(N5CCC(S(=O)(C)=O)CC5)=CC=C4)C(C)=N3)=N2)C=C1.Cl
Molecular Weight (g/mol) 599.02
Formula Weight 599.02
Percent Purity 98.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.