Learn More
Invitrogen™ PTPRN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592949
Description
Immunogen sequence: STGHMILAYM EDHLRNRDRL AKEWQALCAY QAEPNTCATA QGEGNIKKNR HPDFLPYDHA RIKLKVESSP SRSDYINASP IIEHDPRMPA YIATQGPLSH TIADFWQMVW ESGCTVIVML TPLVEDGVKQ CDRYWPDEGA SLYHVYEVNL VSEHIWCEDF LVRSFYLKNV QTQETRTLTQ FHFLSWPAEG TPASTRPLLD FRRKVNKCYR GRSCPIIVHC SDGAGRTGTY ILIDMVLNRM AKGVKEIDIA ATLEHVRDQR PGLVRSKDQF EFALTAVAEE VNAILKALPQ; Positive Samples: 293T, NIH/3T3, Raji, Mouse brain, Rat brian; Cellular Location: Cell junction, Cell membrane, Cell projection, Cytoplasmic vesicle, Endosome, Membrane, Perikaryon, Single-pass type I membrane protein, axon, secretory vesicle membrane, synapse.
IA-2 (insulinoma-associated protein 2, ICA512, PTPRN) is a member of the transmembrane protein tyrosine phosphatase (PTP) family located in secretory granules of neuroendocrine cells. The IA-2 protein is 979 amino acids in length and consists of a luminal domain, transmembrane domain, and cytoplasmic domain. Although a member of the PTP family, IA-2 lacks phosphatase activity with known substrates due to amino acid substitutions at critical sites in its PTP domain. Two paralog RPTPs, IA-2 and IA-2beta were identified as major autoantigens in type-1 diabetes mellitus. On granule exocytosis, the IA-2 cytoplasmic domain is cleaved and the resulting cytosolic fragment moves into the nucleus where it enhances the levels of phosphorylated STAT5 and STAT3, thereby inducing insulin gene transcription and granule biogenesis. IA-2 signaling enhances pancreatic beta-cell proliferation by regulating cyclins D through STATs.
Specifications
| PTPRN | |
| Polyclonal | |
| Unconjugated | |
| PTPRN | |
| 105 kDa islet cell antigen; BEM-3; brain-enriched membrane-associated protein tyrosine phosphatase; IA2; IA-2; IA-2/PTP; ICA 512; ICA105; ICA3; ICA512; ICA512-CCF; ICA512-cleaved cytosolic fragment; ICA512-N-terminal fragment; ICA512-NTF; ICA512-TMF; ICA512-transmembrane fragment; islet cell antigen 2; islet cell antigen 512; Islet cell autoantigen 3; mIA-A; mKIAA4064; protein tyrosine phosphatase, receptor type N; protein tyrosine phosphatase, receptor type, N; protein tyrosine phosphatase, receptor-type, N; protein tyrosine phosphatase-like N; PTP IA-2; PTPLP; Ptprn; receptor-type tyrosine-protein phosphatase-like N; R-PTP-N | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 116660, 19275, 5798 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.77 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16849, Q60673, Q63259 | |
| PTPRN | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.