missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IA-2/PTPRN Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-00134
This item is not returnable.
View return policy
Description
IA-2/PTPRN Polyclonal specifically detects IA-2/PTPRN in Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| IA-2/PTPRN | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CA 512, EC 3.1.3.48, FLJ16131, IA2, IA-2, IA-2/PTP, ICA 512, ICA3, ICA512, islet cell antigen 2, Islet cell antigen 512, Islet cell autoantigen 3, protein tyrosine phosphatase, receptor type, N, protein tyrosine phosphatase-like N, PTP IA-2, receptor-type tyrosine-protein phosphatase-like N, R-PTP-N | |
| Goat | |
| 106 kDa | |
| 0.1 mg | |
| Immune System Diseases, Immunology, Lipid and Metabolism | |
| 5798 | |
| Mouse | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 1.5 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| Q16849 | |
| PTPRN | |
| Peptide with sequence C-QGEGNIKKNRHPD corresponding to internal region according to NP_002837.1. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse. Expected from sequence similarity: Human and Rat. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction