missing translation for 'onlineSavingsMsg'
Learn More

Brady™ i4311 Industrial Label Printer with Brady Workstation and V2200 Barcode Reader Kit

Product Code. 32215715 Shop All Brady Products
Change view
Click to view available options
Connector Types:
Wired
Wired, Bluetooth, Wi-Fi
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
32215715 Wired 1 set
32215711 Wired, Bluetooth, Wi-Fi 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32215715 Supplier Brady™ Supplier No. 325205
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Optimize your operations with an industrial label printer and barcode reader system that streamlines workflows.

  • Total system mobility: Easily transport the lightweight printer and barcode reader together
  • Easy scan and print: Instantly capture data, then print the 6.35 mm to 101.60 mm (0.25 in. to 4 in.) labels you need
  • Continuous uptime: The printer's all-day battery powers the entire system, supporting data capture and printing for up to 5,000 labels per charge
  • Engineered tough: The durable i4311 and the V2200 reader form a reliable solution built for industrial environments
  • Seamless data flow: Connect the V2200 barcode reader via USB-C or USB-A for quick data transfer
  • Fast deployment: Get the complete system running quickly with a simple barcode reader configuration tool and dedicated technical support
  • Software included: Brady Workstation Design & Print Pro helps you create and print using data imports, sequencing, barcoding and more

Specifications

Product Type Printer
Color Blue
Display Type Color LCD Touchscreen
Material Plastic
Print Technology Thermal Transfer
Length (English) 13 in.
Width (English) 9 in.
Length (Metric) 330.20 mm
Width (Metric) 228.60 mm
Height (English) 9.5 in.
Height (Metric) 241.30 mm
Dimensions (L x W x H) 330.20 x 228.60 x 241.30 mm (13 x 9 x 9.5 in.)
Color Capability Monocolor
Display Size 177.80 mm (7 in.)
Includes The i4311 printer ships in a carton box and contains a Li-Ion Battery Pack, an AC Adapter Kit, a USB-C Cord, a Quick Start Guide, a Cleaning Kit, a Mounting bracket, a Brady Workstation Design and Print Pro version and a V2200 Barcode Reader.
Languages Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English (US), Estonian, Finnish, French (Canada), French, German, Hungarian, Italian, Korean, Norwegian, Polish, Portuguese (Brazil), Portuguese, Romanian, Russian, Spanish (Mexico), Spanish, Swedish, Thai, Turkish
Memory 1 GB RAM, 8 GB Flash
Model I4311-V2200-BWS
Print Speed 101.60 mm/sec. (4 in./sec.)
Resolution 300 dpi
Software Type Brady Workstation, Express Labels Mobile App, Windows based driver for 3rd party software use, Software development kit, Brady LINK360™
Connector Types Wired
Operating System Windows™ 11, Windows™ 10, Windows™ 7, Windows™ 8, Windows™ Server 2012, Windows™ Server 2012 R2, Windows™ Server 2016
Weight (English) 12.9 lb.
Weight (Metric) 5.85 kg
Certifications/Compliance CE
For Use With (Application) Access Control, All-Purpose, Asset tracking, Barcode labeling, Circuit board labeling, Cold temperature labeling, Component and equipment labeling, Data and telecommunications labeling, Electrical labeling, Equipment, Exit and egress marking, Facility identification, Floor marking, Food and Beverage Process labeling, Food Handling, General identification, GHS and Chemical Labels, Hazard communication identification, Hazardous Material Labeling, Inspection tagging, Inventory and inspection labeling, Laboratory labeling, Lean and 5S labeling, Lockout tagout, Machinery, Maintenance, Pipe and valve marking, Safety identification, Security identification, Solar Panel labeling, Tamper Indicating Labels, Temperature indicating labeling, Temporary labeling, Traffic Control, Warehouse marking, Wire and cable labeling
Quantity 1
Warranty 1 Year
Power Supply Details Rechargeable Battery, AC Adaptor, USB
Keyboard Type None
Plug Type US, UK, AU, EU
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.