missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Taq DNA Polymerase, native, w/W1

Product Code. 10063172
Click to view available options
Quantity:
100 Unit(s)
500 Unit(s)
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10063172

Brand: Invitrogen™ 18038034

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Taq DNA Polymerase, native, w/W1

Taq DNA Polymerase is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer. The 1% solution of the detergent W-1 that is included may improve the thermostability of the enzyme when added at a final concentration of 0.05% (v/v).

Taq DNA Polymerase consists of a single polypeptide with a molecular weight of 94 kDa. It has a 5´→3´ DNA polymerase activity and a 5´→3´ exonuclease activity (see figure). With Taq DNA Polymerase, you get:

  • Your choice of recombinant or native enzyme
  • Amplification of PCR products up to 5 kb in size
  • An enzyme that is licensed and qualified for PCR

    Applications
    Amplification of DNA from complex genomic, viral, and plasmid templates, RT-PCR, sequencing ssDNA, and cycle sequencing

    Source
    Native enzyme is purified from Thermus aquaticus YT1. Unit definition
    One unit of Taq DNA Polymerase is the amount of enzyme required to incorporate 10 nmoles of deoxyribonucleotide into DNA in 30 min at 74°C.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 5 U/μL
    Content And Storage –20°C
    Description Taq DNA Polymerase, 100 Unit, 5μL Concentration, Liquid, Colorless, 3'-A Overhang, Taq DNA Polymerase, 1X Low GC-Rich PCR Performance, Standard, Reaction Speed, Separate Components Reaction Format
    Form Liquid
    Quantity 100 Unit(s)
    For Use With (Application) Standard PCR
    Overhang 3'-A
    Includes Taq DNA polymerase (20μL), 10X PCR buffer without magnesium (1.25mL), 50mM Magnesium chloride (1mL), 1% W-1 (500μL)
    Source Thermus aquaticus YT1

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.