missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ mirVana™ miRNA Inhibitor, let-7c positive control

Product Code. 13315384
Change view
Click to view available options
Quantity:
20 nmol
5 nmol
Unit Size:
20nmol
5nmol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13315384 5 nmol 5nmol
13325384 20 nmol 20nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13315384 Supplier Invitrogen™ Supplier No. 4464080
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.

miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.

We have now released the second generation of miRNA inhibitors, mirVana™ miRNA Inhibitors, which:

  • Display better potency than lead competitor's product
  • Offer maximum and consistent effect in vitro at low miRNA inhibitor concentration
  • Target the most updated Sanger content
  • Can be used in vitro and in vivo, offering consistency throughout your entire research project
  • Are non-toxic

Positive and negative controls are also available for use along with mirVana™ miRNA Inhibitors in miRNA down-regulation experiments.

mirVana™ miRNA Inhibitor let-7c Positive Control

The mirVana™ miRNA Inhibitor let-7c Positive Control is intended for use as a positive control for mammalian cell culture experiments using mirVana™ miRNA Inhibitors. Endogenous let-7c miRNA has been shown to negatively regulate HMGA2 mRNA in cultured cells. HMGA2 is a ubiquitously expressed, nonhistone, chromatin protein that can modulate gene expression through changes in chromatin architecture. let-7c miRNA down-regulates levels of HMGA2 mRNA. When transfected into human and mouse cell lines, let-7c miRNA inhibitor blocks endogenous let-7c miRNA, resulting in increased levels of HMGA2 mRNA. Thus, let-7c miRNA inhibitor activity can be monitored in human or mouse cells using real-time reverse transcription PCR (RT-PCR) to detect HMGA2 mRNA.

First Generation Inhibitor Controls

Our first-generation miRNA inhibitors (and controls), Ambion™ Anti-miR™ miRNA Inhibitors, have lower specificity and potency, plus a lower strand specificity compared to the new mirVana™ miRNA Inhibitors. These first-generation inhibitors are recommended for completing ongoing in vitro miRNA studies. For new studies or in vivo studies, we highly recommend our new mirVana™ miRNA Inhibitors and their corresponding controls.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at or below -20°C. Do not store in a frost-free freezer.
Control Type Positive Control
RNAi Type miRNA Inhibitor
For Use With (Application) miRNA & Non-Coding RNA Analysis
Label or Dye Unconjugated
Product Line mirVana
Product Type miRNA Inhibitor
Purification Method Purified
Quantity 5 nmol
Shipping Condition Room Temperature
How can I quantify endogenous miRNA inactivation by mirVana inhibitors?

Using TaqMan miRNA assays, you can measure the level of miRNA of interest. If the miRNA is bound to the synthetic anti-miRs and/or mirVana inhibitors transfected, the duplex should not be detected, indicating efficient miRNA inactivation. Please note, this only works when total RNA sample isolated is not heat denatured when RT primer is added, as heat denaturation will result in complex dissociation and subsequent detection of microRNA by TaqMan miRNA assay even when cells have been transfected properly. Please see the protocol for the TaqMan MicroRNA Cells-to-CT kit (Cat. No. 4391848) for more details.

How do you introduce mirVana miRNA mimics or inhibitors into cells?

Mimics and inhibitors can be transfected or electroporated into cells using parameters similar to those used for siRNA.

Using mirVana miRNA Mimics- For the following applications, use a cell line that expresses relatively low levels of the mature miRNA of interest.
- Cell density at the time of transfection. In general, 30-70% confluency is recommended.
- Transfection method (traditional or reverse transfection).
- Length of exposure of cells to transfection agent-RNA oligonucleotides complexes.
For many miRNA mimics, maximal activity is achieved after 24 hours, but biological assays may be performed 24-72 hours posttransfection. If you maintain your transfected cells longer than 24 hours, we suggest replacing the existing medium with fresh medium at 24 hours posttransfection, for greater viability of the cells.

Using mirVana miRNA Inhibitors to analyze endogenous miRNA function
For the following applications, use a cell line that expresses relatively high levels of the mature miRNA of interest.
General transfection starting points
mirVana miRNA Mimics and Inhibitors typically work best when transfected at a final concentration of 3-30 nM. Optimization experiments might include a concentration range of 1-100 nM. To increase accuracy and reproducibility when preparing transfection complexes, prepare a dilution of your stock oligonucleotides, and pipet a higher volume of diluted stock.

What is the turnaround time for your mirVana miRNA Mimic and Inhibitors?

The turnaround time varies depending on the level of purification and whether it is a custom order or not. Please see the approximate timetable below:

Standard purification (5 and 20 nmol), 4 business days
HPLC/in vivo ready (250 nmol), 15 business days
Custom mirVana Mimics and Inhibitors, 15 business days

I would like to create a custom mirVana mimic and inhibitor. Can I do this?

Yes, please provide us with your unpublished miRNA sequence and we can synthesize a corresponding mirVana mimic or inhibitor by clicking on this link (www5.invitrogen.com/custom-genomic-products/tools/mirna/).

Can I get the sequence for the mirVana Mimics and Inhibitors negative control?

Negative control sequences require a confidential disclosure agreement while target sequences require prior proof of purchase for release.

How should I store my mirVana mimics and inhibitors?

Store the dried oligonucleotides in a non-frost-free freezer at or below -20 degrees C. Store resuspended oligonucleotides at or below -20 degrees C for extended periods (up to 1 year), and they can undergo up to 50 freeze/thaw cycles without significant degradation. We recommend aliquoting the resuspended miRNA to prevent contamination. Storage at -80 degrees C is fine but not necessary. Working stock solutions can be stored at 4 degrees C for up to 7 days before use.

Do you offer mirVana miRNA Inhibitor controls?

Yes, we offer both positive and negative controls for our mirVana miRNA inhibitors. The mirVana miRNA Inhibitor let-7c Positive Control is intended for use as a positive control for mammalian cell culture experiments using mirVana miRNA Inhibitors (Cat. No. 4464080, 4464081). Endogenous let-7c miRNA has been shown to negatively regulate HMGA2 mRNA in cultured cells. HMGA2 is a ubiquitously expressed, non-histone, chromatin protein that can modulate gene expression through changes in chromatin architecture. let-7c miRNA down-regulates levels of HMGA2 mRNA. When transfected into human and mouse cell lines, let-7c miRNA inhibitor blocks endogenous let-7c miRNA, resulting in increased levels of HMGA2 mRNA. Thus, let-7c miRNA inhibitor activity can be monitored in human or mouse cells using real-time reverse transcription PCR (RT-PCR) to detect HMGA2 mRNA.

mirVana miRNA Inhibitor Negative Control #1 is intended for use as a negative control for experiments using mirVana miRNA Inhibitors (Cat. No. 4464076, 4464077, 4464078, 4464079). Transfect this negative control in parallel with your positive control (such as let-7c Positive Control) and experimental mirVana miRNA Inhibitors. Use target expression from negative control-transfected samples as a baseline for evaluation of the effect of the control and experimental miRNA inhibitor on target gene expression.

Do you offer mirVana miRNA mimic controls?

Yes, we offer both positive and negative controls for our mirVana miRNA mimics. The mirVana miRNA Mimic miR-1 Positive Control (Cat. No. 4464062, 4462063, 4462064, 2262065) is designed to mimic mature miR-1 miRNA after delivery to mammalian cells. miR-1 is associated with down-regulation of many genes. In particular, miR-1-mediated down-regulation of Protein Tyrosine Kinase 9 (PTK9) has been shown to occur at the mRNA level. miR-1 Positive Control is intended for use as a positive control for transfection experiments using mirVana miRNA Mimics. Monitor activity of miR-1 Positive Control through its gene silencing of PTK9, using real-time reverse transcription-PCR to detect PTK9 mRNA.
mirVana miRNA Mimic Negative Control #1 (4464058, 4464059, 4464060, 4464061) is designed to use as a negative control for experiments using mirVana miRNA Mimics. Transfect the Negative Control using the same methodology as for your positive control (such as miR-1 Positive Control) and experimental mirVana miRNA Mimics. Use target gene expression from Negative Control-transfected samples as a baseline for evaluation of the effect of the control and experimental miRNA mimic on target gene expression. mirVana miRNA Mimic Negative Control #1 is a random sequence miRNA mimic molecule that has been extensively tested in human cell lines and tissues and validated to not produce identifiable effects on known miRNA function.

What is the molecular weight for the mirVana miRNA mimics and mirVana miRNA inhibitors?

The average molecular weight for mirVana miRNA mimics is 14,000 g/mol (where 1 nmol = 14 µg), and 11,000 - 12,000 g/mol (where 1 nmol = 11 - 12 µg) for mirVana miRNA inhibitors. The exact molecular weights are proprietary.

Where can I find more information about published miRNA sequences and annotation?

Please visit miRBase: The microRNA database (http://www.mirbase.org/).

I would like to order a mimic or inhibitor with a 5' label. Can you do this?

We can label our Pre-miR and Anti-miR miRNA, but do not offer labeled mirVana Mimics and inhibitors. 5' modifications include Alexa Fluor 555, Alexa Fluor 647, FAM, and Cy3. Request a quote here (http://www.thermofisher.com/us/en/home/global/forms/custom-sirna-mirna-quote-request.html).

The miRNA sequence I need to target is not in the miRBase database. Can I order a custom mimic or inhibitor?

Yes, submit your unpublished miRNA sequences and/or order custom modified miRNA here (http://www.thermofisher.com/order/custom-genomic-products/tools/mirna/).

What is the difference between Invitrogen Anti-miR inhibitors & Invitrogen Pre-miR miRNA Precursors and mirVana miRNA mimics and inhibitors?

Invitrogen Anti-miR inhibitors & Invitrogen Pre-miR miRNA Precursors are our first generation mimic and inhibitors. Invitrogen Anti-miR miRNA Inhibitors are chemically modified, single stranded nucleic acids designed to specifically bind and inhibit endogenous microRNA (miRNA) molecules. These are ready to use inhibitors that can be introduced into cells using transfection or electroporation. Pre-designed inhibitors have been created for all miRNAs listed in miRBase version 15.0.
Invitrogen Pre-miR miRNA Precursors are small, chemically modified double-stranded RNA molecules designed to mimic endogenous mature miRNAs. These ready-to-use miRNA mimics, which are similar to, but not identical to siRNAs, can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. Please note, Pre-miR miRNA Precursors are not hairpin constructs and should not be confused with pre-miRNAs.
mirVana miRNA mimics and inhibitors are our second generation of mimics and inhibitors. mirVana miRNA mimics are small, chemically double-stranded RNAs that mimic endogenous mature miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity. They have enhanced specificity over the first generation mimics, are delivered as HPLC purified, and ready to transfect.
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity. They have enhanced potency over our first generation inhibitors. Pre-designed inhibitors were created for all miRNAs listed in miRBase version 19.0. They come delivered as HPLC purified, and ready to transfect. Learn more about mirVana miRNA mimics and inhibitors through this video (https://www.youtube.com/watch?feature=player_embedded&v=Q3MO1lmT0kw). Both mimics and inhibitors are available individually or as libraries.
They are compatible with both in vitro and in vivo applications, and have been validated with Lipofectamine RNAiMAX Transfection Reagent for use in cell-based systems, and with Invivofectamine 3.0 Transfection Reagent for in vivo delivery. These oligonucleotides are nontoxic and do not induce an immune response in the animal models tested. In vivo-ready mirVana miRNA mimics and inhibitors have been purified by HPLC and dialysis and are ready for immediate in vivo use. For all library inquiries, please email: rnailibraries@lifetech.com.

What is the difference between siRNA and miRNA?

siRNA mediates degradation of a target mRNA molecule in a sequence-specific manner, while miRNA suppresses translation without cleaving mRNA.

What is miRNA?

miRNA (microRNAs) are short, highly conserved small noncoding RNA molecules naturally occurring in the genomes of plants and animals. miRNAs are single stranded, 18-25 nucleotides long and regulate posttranscriptional mRNA expression, typically by binding to the 3' untranslated region (3'-UTR) of the complementary mRNA sequence, resulting in translational repression and gene silencing. Studies have shown that thousands of human protein-coding genes are regulated by miRNAs, indicating that miRNAs are “master regulators” of many important biological processes.

Are mirVana miRNA Mimics delivered in buffer or dry/lyophilized?

These products are delivered dry lyophilized.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.