missing translation for 'onlineSavingsMsg'
Learn More

I-CBP112, MedChemExpress

Product Code. 16477798
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477798 10 mg 10mg
16457636 100 mg 100mg
16443757 50 mg 50mg
16431829 1 mg 1mg
16410268 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16477798 Supplier Medchem Express Supplier No. HY19541

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress I-CBP112 is a specific and potent acetyl-lysine competitive protein-protein interaction inhibitor, that inhibits the CBP/p300 bromodomains, enhances acetylation by p300.

I-CBP112 is a specific and potent acetyl-lysine competitive protein-protein interaction inhibitor, that inhibits the CBP/p300 bromodomains, enhances acetylation by p300.

Specifications

Chemical Name or Material I-CBP112
CAS 1640282-31-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H36N2O5
Quantity 10 mg
Solubility Information DMSO : ≥ 32 mg/mL (68.29 mM)
SMILES COC1=C(OC)C=C(C2=CC(OC[C@H]3CCCN(C)C3)=C(OCCN(C(CC)=O)C4)C4=C2)C=C1
Molecular Weight (g/mol) 468.59
Formula Weight 468.59
Percent Purity 98.46%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.