missing translation for 'onlineSavingsMsg'
Learn More

I-BET282, MedChemExpress

Product Code. 16473837
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473837 50 mg 50mg
16447706 100 mg 100mg
16440358 5 mg 5mg
16407898 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473837 Supplier Medchem Express Supplier No. HY19760

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress I-BET282 is a pan-inhibitor of all eight BET bromodomains, and selectivity over other representative bromodomain-containing proteins. I-BET282 shows pIC50s ranging 6.4-7.7 for BRD2 (BD1/BD2), BRD2 (BD1/BD), BRD3 (BD1/BD), and BRD4 (BD1/BD).

I-BET282 is a pan-inhibitor of all eight BET bromodomains, and selectivity over other representative bromodomain-containing proteins. I-BET282 shows pIC50s ranging 6.4-7.7 for BRD2 (BD1/BD2), BRD2 (BD1/BD), BRD3 (BD1/BD), and BRD4 (BD1/BD).

Specifications

Chemical Name or Material I-BET282
CAS 1422554-34-4
Color White
Purity Grade Notes Research
Molecular Formula C25H30N4O4
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (221.96 mM; Need ultrasonic)
SMILES COC1=C(C2=C(C)ON=C2C)C=C(N=CC3=C4N(C(C5CCOCC5)=N3)[C@@H](COC)C)C4=C1
Molecular Weight (g/mol) 450.53
Formula Weight 450.53
Percent Purity 99.12%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.