missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies I-BET151

Product Code. 17170262
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17170262 50 mg 50mg
17160262 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17170262 Supplier STEMCELL Technologies Supplier No. 73714

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

BET family inhibitor; Inhibits BRD2, BRD3, and BRD4

  • I-BET151 is an inhibitor of bromodomain and extra terminal (BET) family proteins
  • BET proteins recognize acetylated lysine residues via their 2 bromodomains (Gallenkamp et al.) I-BET151 inhibits BRD2, BRD3, and BRD4 with IC50 values of 0.5, 0.25, and 0.79 μM, respectively (Kline et al.; Vidler et al.; Hewings et al.; Dawson et al. 2012)
  • Enhances reprogramming of mouse fibroblasts to neurons, in combination with ISX-9, Forskolin, and CHIR99021 (Li et al.)
  • Induces early cell cycle arrest and apoptosis in human and mouse MLL-fusion leukemia cell lines by blocking transcription of key genes including BCL2, C-MYC, and CDK6 (Dawson et al.) 2011)

Specifications

CAS 1300031-49-5
Synonym GSK1210151A
Quantity 50 mg
Molecular Formula C23H21N5O3
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.