Learn More
BUCHI I-80 Controller for Rotavapor R-80
Product Code. p-200020849 Shop All Buchi Products
Click to view available options
For Use With (Equipment):
Rotavapor R-80 House Vacuum
Rotavapor R-80 V-80/180
Includes:
Communication Cable, Holder
Power Pack
Woulff Bottle, Communication Cable, Tubing
Unit Size:
Each
Product Code. 30000637
Brand: BUCHI 11I0800
This item requires a price update prior to being ordered.
Request a Price for this item here.
The I-80 is an easy-to-use interface for conveniently regulating your vacuum manually. Increase control of the evaporation process by setting the pump speed and adjusting the manometer.
  • Functional design fulfills essential needs of vacuum regulation
  • Saves resources thanks to control of vacuum pump
  • High longevity due to robust and durable design
  • User-friendly and intuitive handling due to self-explaining user interface
  • Easy and accurate vacuum regulation
  • Relevant information is displayed at a glance
  • High reliability due to centralized control of major process parameters
  • Full process control thanks to parameter monitoring
  • Proven technology for major applications
Specifications
Type Controller Interface
For Use With (Equipment) Rotavapor R-80 House Vacuum
Includes Woulff Bottle, Communication Cable, Tubing