missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ I 432
Description
I 432 is a TMPRSS2 (transmembrane serine proteinase 2) inhibitor (Ki =0.9 nM). It blocks influenza virus propagation in airway epithelial cells. In IPEC-J2 pig jejunal epithelial cells, I 432 (50 μM) inhibits TMPRSS2 without affecting cell viability after 48 h treatment.
Specifications
Specifications
| Chemical Name or Material | 3-[(2 S)-3-[4-(2-Aminoethyl)-1-piperidinyl]-2-[[(2 ',4 '-dichloro[1,1 '-biphenyl]-3-yl)sulfonyl]amino]-3-oxopropyl]benzenecarboximidamide ditrifluoroacetate |
| CAS | 1268874-09-4 |
| Quantity | 5 mg |
| Target | TMPRSS2 |
| Molecular Formula | C29H33Cl2N5O3S0.2 CF3CO2H |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?