Learn More
BUCHI I-180 Controller for Rotavapor R-80
Product Code. p-200020850 Shop All Buchi Products
Click to view available options
For Use With (Equipment):
Rotavapor R-80 I-100
Rotavapor R-80 V-80/180
Rotavapor R-80 V-80/180 House Vacuum
Includes:
Adapter Cable for I-180, I-100 Power Supply
Communication Cable, Holder
Power Pack
Valve Unit, Woulff Bottle, Communication Cable, Tubing
Unit Size:
Each
Product Code. 30000610
Brand: BUCHI 11I1802
This item requires a price update prior to being ordered.
Request a Price for this item here.
The I-180 is an easy-to-use interface for conveniently regulating your vacuum automatically. Increase control of the evaporation process by setting the desired vacuum and accessing other modes of operation.
  • Functional design fulfills essential needs of vacuum regulation
  • Saves resources thanks to control of vacuum pump
  • High longevity due to robust and durable design
  • User-friendly and intuitive handling due to self-explaining user interface
  • Easy and accurate vacuum regulation
  • Relevant information is displayed at a glance
  • High reliability due to centralized control of major process parameters
  • Full process control thanks to parameter monitoring
  • Proven technology for major applications
Specifications
Type Controller Interface
For Use With (Equipment) Rotavapor R-80 V-80/180
Includes Power Pack