missing translation for 'onlineSavingsMsg'
Learn More

Hypromellose, TRC

Product Code. 30134498
Change view
Click to view available options
Quantity:
10 g
100 g
250 g
Unit Size:
100g
10g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134498 100 g 100g
30138237 250 g 250g
30040871 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134498 Supplier Toronto Research Chemicals Supplier No. TRCH998600100G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-65-3
Synonym 2-Hydroxypropyl Methyl Cellulose, 2-Hydroxypropyl Methyl Cellulose Ether, 40US, 60HD20000, 60MP4000, 60RT50, 60SH06, 60SH100, 60SH4000, 60SH4000F, 60SH5, 65SH-4000, 65SH5, 90SH100, 90SH100000, 90SH15000S, 90SH400, Accel R 100, BN 4, Benecel 324, Benecel 363, Benecel 424, Benecel K 35M, Benecel MP 3, Benecel MP 363C, Benecel MP 824, Benecel MP 843, Benecel MP 9, Benecel MP 943, Benecel MP 943W, Celacol 15000DS, Celacol HPM 15000DS, Celacol HPM 450, Celacol HPM 5000, Cellulose hydroxypropyl methyl ether, Cesca HPC 50, Courlose HPM, Culminal 1034, Fortefiber, Hydroxypropyl methyl cellulose, Hydroxypropyl methyl cellulose ether, Hypromelloc E 5, Hypromellose, Hypromellose 2208, Hypromellose 2910, Hypromellose E 15, Hypromellose E 5, Hypromellose K 100MCR, Magimix, Marpolose, Mecellose, Methocel, Methofas, Methyl hydroxypropyl cellulose, Metolose, Neovisco MC HM 4000, Neovisco MC RM 30000, Nisso L, Nisso-SL, Nisso-SSL, Noigel 0215H, Onoda Elcon, Opadry OY 29020 clear, Opadry OY 7240, Opadry YS 1-7472, PM 450, PM 4500, PMB 40H, PMC 40HS, Pharmacoat 603, Pharmacoat 606, Pharmacoat 615, Pharmacoat 645, Pharmacoat 904, Pharmacoat HPMC 615, Quali-V, R 40, R 40 (cellulose derivative), SB 4, SB 4 (cellulose derivative), SGA 16M, Sepifilm LP, Sepifilm LP 010, Spectracel 5FG, Synchron, Synchron-Oral, TC 5, TC 5 (cellulose derivative), TC 5E, TC 5EW, TC 5MW, TC 5R, TC 5RW, TC 5S, TC-5 RG, Tegocel, Tegocel Fluid HPM 50, Tegocel HPM 4000, Tegocel HPM 50, Vcaps, Vcaps Plus, Viscontran MHPC 50, Viscontran MHPC 6000, Walocel HM, Walocel HM 100PA2208FG, Walocel HM 50PA2910, Walocel HM 6PA2910, Walocel MK 15000PF, Walocel MK 3.000PF, Walocel MK 400PF, Walocel MW 60GA

Specifications

Quantity 100 g
Chemical Name or Material Hypromellose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.