missing translation for 'onlineSavingsMsg'
Learn More

Hypoxanthine, MedChemExpress

Product Code. 16458976
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458976 100 mg 100mg
16454747 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16458976 Supplier Medchem Express Supplier No. HYN0091
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hypoxanthine, a purine derivative, is a potential free radical generator and could be used as an indicator of hypoxia.

Hypoxanthine, a purine derivative, is a potential free radical generator and could be used as an indicator of hypoxia.

Specifications

Chemical Name or Material Hypoxanthine
CAS 68-94-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C5H4N4O
Quantity 100 mg
Synonym Purin-6-ol Sarcine
Solubility Information DMSO : 10 mg/mL (73.47 mM; Need ultrasonic)
SMILES O=C1NC=NC2=C1N=CN2
Molecular Weight (g/mol) 136.11
Formula Weight 136.11
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.