missing translation for 'onlineSavingsMsg'
Learn More

Hypoxanthine, 99%

Product Code. 11460373
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460373 5 g 5g
11470373 25 g 25g
11480373 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460373 Supplier Thermo Scientific Alfa Aesar Supplier No. A11481.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 68-94-0 | C5H4N4O | 136.11 g/mol

Hypoxanthine, is used as a marker for energy perturbation in hypoxia/ ischemia. It has been also used in DNA studies to investigate the destabilizing effect it has on DNA duplexes containing hypoxanthine as a base, in a gas phase versus a liquid phase. Hypoxanthine has also been used in studies along with 8-oxoguanine nucleotides to investigate their interaction with human DNA pol A (DNA polymerase alpha) and DNA polymerase I from Bacillus stearothermophilus. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hypoxanthine, is used as a marker for energy perturbation in hypoxia/ ischemia. It has been also used in DNA studies to investigate the destabilizing effect it has on DNA duplexes containing hypoxanthine as a base, in a gas phase versus a liquid phase. Hypoxanthine has also been used in studies along with 8-oxoguanine nucleotides to investigate their interaction with human DNA pol å (DNA polymerase alpha) and DNA polymerase I from Bacillus stearothermophilus. It is also used as a pharmaceutical intermediate.

Solubility
It does not mix appreciably with water (1:1400). Small octahedra crystals from water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents. Stable under recommended storage conditions. Sterilize by filtration through a 0.22 µm filter.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-94-0
Molecular Formula C5H4N4O
Molecular Weight (g/mol) 136.11
MDL Number MFCD00005725
InChI Key FDGQSTZJBFJUBT-UHFFFAOYSA-N
Synonym hypoxanthine, 6-hydroxypurine, 9h-purin-6-ol, sarkin, 6-oxopurine, sarcine, sarkine, 3h-purin-6-ol, 7h-purin-6-ol, purin-6 1h-one
PubChem CID 790
ChEBI CHEBI:17368
IUPAC Name 3,7-dihydropurin-6-one
SMILES O=C1N=CNC2=C1NC=N2

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 5811
Merck Index 14,4869
Solubility Information It does not mix appreciably with water (1:1400). Small octahedra crystals from water.
Formula Weight 136.11
Percent Purity 99%
Chemical Name or Material Hypoxanthine
compliance-icons
Product Identifier
  • Hypoxanthine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Hypoxanthine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.