missing translation for 'onlineSavingsMsg'
Learn More

Hypoxanthine-13C2,15N (Mixture), TRC

Product Code. 30401918
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401918 10 mg 10mg
30391484 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401918 Supplier Toronto Research Chemicals Supplier No. TRCH99850310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246820-04-1
Molecular Formula C313C2H4N315NO
Molecular Weight (g/mol) 139.09
Synonym 1,9-Dihydro-6H-purin-6-one-13C2,15N, 3H-Purin-6-ol-13C2,15N, 6-Hydroxy-1H-purine-13C2,15N, 6-Hydroxypurine-13C2,15N, 6-Oxopurine-13C2,15N, Hypoxanthine Enol-13C2,15N, NSC 129419-13C2,15N, NSC 14665-13C2,15N, Purin-6-ol-13C2,15N, Sarcine-13C2,15N, Sarkin-13C2,15N, Sarkine-13C2,15N
SMILES O=C1C([15NH]C=N2)=[13C]2N[13CH]=N1.O=C3C([15NH][13CH]=N4)=[13C]4NC=N3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/2C5H4N4O/c2*10-5-3-4(7-1-6-3)8-2-9-5/h2*1-2H,(H2,6,7,8,9,10)/i2+1,4+1,6+1;1+1,4+1,6+1
Formula Weight 139.04
Chemical Name or Material Hypoxanthine 4,2 or 8-13C2,7-15N (mixture of 13C sites)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.