missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hypotaurine, MedChemExpress
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Hypotaurine (2-aminoethanesulfinic acid), an intermediate in taurine biosynthesis from cysteine in astrocytes, is an endogenous inhibitory amino acid of the glycine receptor. Antioxidant.
Specifications
Specifications
| Chemical Name or Material | Hypotaurine |
| Physical Form | Powder |
| CAS | 300-84-5 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C2H7NO2S |
| For Use With (Application) | Neuroscience-Neurodegeneration |
| Synonym | 2-Aminoethanesulfinic acid |
| Solubility Information | H2O : 125 mg/mL (1145.21 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction