missing translation for 'onlineSavingsMsg'
Learn More

Hypophosphorous Acid Solution, Honeywell

Product Code. p-7071873 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 15621970

Brand: Honeywell Fluka 214906100ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6303-21-5
Molecular Formula H3O2P
Molecular Weight (g/mol) 66.00
MDL Number MFCD02183592
InChI Key ACVYVLVWPXVTIT-UHFFFAOYSA-N
Synonym hypophosphite ion, hypophosphorus acid, phosphinic acid, ion 1-, hydroxyphosphanone, oxo-, e5-phosphanol, hydroxidooxidophosphorus, perfluoroalkyl c6-c12 phosphonic acid, diperfluoroalkyl c6-c12 phosphinic acid, hydroxy-, e5-phosphanone, perfluoro-c6-c12 ethyl hydrogen phosphate
PubChem CID 22497
ChEBI CHEBI:36363
IUPAC Name phosphinic acid
SMILES O[PH2]=O

Specifications

Concentration 50wt. % in H2O
Density 1.206g/mL at 25°C
Vapor Pressure 17mmHg (20°C)
Quantity 100 mL
Linear Formula H3PO2
UN Number 3264
Formula Weight 66g/mol
Percent Purity 50% (wt. in H2O)
Chemical Name or Material Hypophosphorous Acid Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.