missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersil™ MOS C8 HPLC Columns

Product Code. 10374373 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
125 mm
150 mm
200 mm
250 mm
Diameter (Metric):
3 mm
4 mm
4.6 mm
Particle Size:
3 μm
5 μm
10 μm
Unit Size:
Each
Pack of 4
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size unitSize
10374373 10 mm 4 mm 5 μm Pack of 4
10322545 50 mm 4.6 mm 5 μm Each
11557461 125 mm 4 mm 3 μm Each
10120793 150 mm 4.6 mm 5 μm Each
10528015 150 mm 4 mm 5 μm Each
10552855 250 mm 4 mm 5 μm Each
10353993 250 mm 4.6 mm 5 μm Each
10392765 250 mm 3 mm 5 μm Each
11527471 200 mm 4.6 mm 10 μm Each
11537471 250 mm 4.6 mm 10 μm Each
10027214 50 mm 4.6 mm 3 μm Each
10640146 150 mm 4.6 mm 3 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 10374373 Supplier Thermo Scientific™ Supplier No. 30205014001
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use as a general-purpose C8 phase for acidic and neutral analytes.

For reliable, affordable analysis of more hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids, and environmental samples, use Thermo Scientific™ Hypersil™ MOS C8 HPLC Columns.

Recognized as an HPLC industry standard, Hypersil stationary phases are proven effective analytical tools. Classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 guidelines.

For separations requiring improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ C8 columns as an alternative.

  • The 10mm design offers maximum protection with minimal increase in retention Fit Thermo Scientific UNIGUARD direct connection and stand alone holders
  • Available in packs of 4

Feature of MOS (C8) Solid Phase

  • Reliable, less retentive phase
TRUSTED_SUSTAINABILITY

Specifications

Column Format Guard Cartridge, 4 Pk.
Diameter (Metric) 4 mm
For Use With Uniguard Direct-Connection and Stand Alone Holder
Length (Metric) 10 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 120 Å
Surface Area 170 m²/g
Temperature 60°C
USP Type L7
Stationary Phase C8
Product Line Hypersil Classical
Packing Material Silica
Carbon Load 6.5%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.