Learn More
Thermo Scientific™ Hypersil™ MOS C8 HPLC Columns
Use as a general-purpose C8 phase for acidic and neutral analytes.
Brand: Thermo Scientific™ 30203-154630
Code : 16
Additional Details : Weight : 0.10000kg
Description
For reliable, affordable analysis of more hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids, and environmental samples, use Thermo Scientific™ Hypersil™ MOS C8 HPLC Columns.
Recognized as an HPLC industry standard, Hypersil stationary phases are proven effective analytical tools. Classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 guidelines.
For separations requiring improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ C8 columns as an alternative.
- Reliable, less retentive phase
- Long column lifetimes, even under basic conditions
- Manufactured under ISO 9001-2000 quality guidelines
- Affordable
Ordering Information:
Other column dimensions are available. Also available in 10μm. Please call Customer Service for more information.
Specifications
Analytical Column | |
HPLC | |
5800 psi (400 bar) | |
3 μm | |
120 Å | |
60°C | |
C8 | |
Spherical, Fully Porous Type A Silica | |
Reversed-phase | |
Reversed Phase |
4.6 mm | |
150 mm | |
Spherical | |
2 to 8 | |
170 m²/g | |
L7 | |
Hypersil Classical | |
6.5% | |
No |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.