missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersil GOLD™ C4 Reversed Phase HPLC Column, 3 μm, 1 mm x 50 mm

Product Code. 10332445 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
50 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10332445 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10332445 Supplier Thermo Scientific™ Supplier No. 25503051030

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Column Format: Analytical Column; Carbon Load: 5%; Pore Size: 175 Å; Column Phase: Reversed Phase

Thermo Scientific™ Hypersil GOLD 3μm C4 Analytical HPLC Columns features lower hydrophobicity than C18 or C8 and are recommended for very hydrophobic analytes. Based on high purity silica, Hypersil GOLD is a state-of-the-art family of columns designed to meet the challenges of modern chromatography.

Similar Selectivity, Less Retention Than C18 or C8 Columns

  • Successful retention and separation of very hydrophobic analytes with this short alkyl chain length, low hydrophobicity column
  • Ideal for analysis of hydrophobic polypeptides and proteins

Superior Performance

  • Highly symmetrical peak shapes 
  • 1.9μm particle size columns improve speed and efficiency

About the Hypersil GOLD Family of HPLC Columns

Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.

  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 3μm particle size for your separation needs
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 1 mm
Flow Rate 0.05 mL/min
For Use With HPLC
Length (Metric) 50 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 3 μm
pH 2 to 8
Pore Size 175 Å
Surface Area 220 m²/g
USP Type L26
Stationary Phase C4
Product Line Hypersil GOLD
Packing Material Silica
Carbon Load 5%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.